ENA333558333 Enamine

2-amino-2-cyclopropylpropanamide hydrochloride

  • MDL number MFCD19982487
  • Cas Number : 1334149-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83146
Availability details for
Quantity
  • Availability
  • Add to Cart