ENA333558352 Enamine

3-methanesulfonylbutan-2-amine hydrochloride

  • MDL number MFCD19982523
  • Cas Number : 1334148-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83434
Availability details for
Quantity
  • Availability
  • Add to Cart