ENA333558357 Enamine

(1H-imidazol-4-yl)methanamine dihydrochloride

  • MDL number MFCD09056757
  • Cas Number : 72631-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82476
Availability details for
Quantity
  • Availability
  • Add to Cart