ENA333558359 Enamine

1-methyl-1H-1,2,3-benzotriazole-5-carbaldehyde

  • MDL number MFCD03407337
  • Cas Number : 499770-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82478

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-82478
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart