ENA333558362 Enamine

1-propyl-1H-indol-6-amine

  • MDL number MFCD11638202
  • Cas Number : 1095532-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82481
Availability details for
Quantity
  • Availability
  • Add to Cart