ENA333558363 Enamine

1-ethyl-1H-indol-6-amine

  • MDL number MFCD11638177
  • Cas Number : 681836-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82482
Availability details for
Quantity
  • Availability
  • Add to Cart