ENA333558366 Enamine

methyl 4-(2-oxopyrrolidin-1-yl)butanoate

  • MDL number MFCD09037920
  • Cas Number : 6740-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82492
Availability details for
Quantity
  • Availability
  • Add to Cart