ENA333558369 Enamine

3-(aminomethyl)-2-methylpentane

  • MDL number MFCD12807248
  • Cas Number : 163628-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82495

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82495
Availability details for
Quantity
  • Availability
  • Add to Cart