ENA333558373 Enamine

4-hydrazinylidene-N,N-dimethyl-1,4-dihydropyrimidin-2-amine

  • Cas Number : 117077-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82500
Availability details for
Quantity
  • Availability
  • Add to Cart