ENA333558375 Enamine

2-(6-amino-1H-indol-1-yl)-N-methylacetamide

  • MDL number MFCD11638187
  • Cas Number : 1096264-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82502
Availability details for
Quantity
  • Availability
  • Add to Cart