ENA333558376 Enamine

1-[1-(4-bromophenyl)cyclopentyl]methanamine

  • MDL number MFCD10689718
  • Cas Number : 626603-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82504
Availability details for
Quantity
  • Availability
  • Add to Cart