ENA333558379 Enamine

1-bromo-4-[(propan-2-yloxy)methyl]benzene

  • MDL number MFCD14687019
  • Cas Number : 98446-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82507

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82507
Availability details for
Quantity
  • Availability
  • Add to Cart