ENA333558383 Enamine

5-amino-1-benzyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD02219534
  • Cas Number : 25784-56-9
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-82311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82311
Availability details for
Quantity
  • Availability
  • Add to Cart