ENA333558387 Enamine

3-(4-fluorobenzoyl)piperidine hydrochloride

  • MDL number MFCD11846283
  • Cas Number : 118412-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82518
Availability details for
Quantity
  • Availability
  • Add to Cart