ENA333558390 Enamine

2-amino-N-benzyl-3-phenylpropanamide hydrochloride

  • MDL number MFCD13562759
  • Cas Number : 34582-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82521
Availability details for
Quantity
  • Availability
  • Add to Cart