ENA333558399 Enamine

5-chloropyridine-3-sulfonyl chloride

  • MDL number MFCD13189042
  • Cas Number : 1060802-18-7
  • Molecular Weight : 212.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-pyridinesulfonyl chloride
Inchi Code 1S/C5H3Cl2NO2S/c6-4-1-5(3-8-2-4)11(7,9)10/h1-3H
Inchi Key FQNDXOOGXYFWHZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-82533

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-82533
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart