ENA333558402 Enamine

1,3-diethyl 2-(3-chlorophenyl)propanedioate

  • MDL number MFCD06205061
  • Cas Number : 93307-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82543

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82543
Availability details for
Quantity
  • Availability
  • Add to Cart