ENA333558407 Enamine

N-methyl-N-(2H-1,2,3,4-tetrazol-5-ylmethyl)benzamide

  • MDL number MFCD19686494
  • Cas Number : 1333806-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136...138
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82548

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82548
Availability details for
Quantity
  • Availability
  • Add to Cart