ENA333558410 Enamine

6-methoxypyridazine-3-carboxylic acid

  • MDL number MFCD09800587
  • Cas Number : 56434-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-163
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82553
Availability details for
Quantity
  • Availability
  • Add to Cart