ENA333558411 Enamine

3-(1H-1,2,4-triazol-5-yl)benzoic acid

  • MDL number MFCD07643205
  • Cas Number : 876715-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82555
Availability details for
Quantity
  • Availability
  • Add to Cart