ENA333558414 Enamine

1-chloro-4-(4-methoxyphenyl)phthalazine

  • MDL number MFCD03030035
  • Cas Number : 128615-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82558

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82558
Availability details for
Quantity
  • Availability
  • Add to Cart