ENA333558418 Enamine

4-(2-methylpropyl)-3-(pyrrolidin-2-yl)-4H-1,2,4-triazole

  • MDL number MFCD16852338
  • Cas Number : 1247132-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82326
Availability details for
Quantity
  • Availability
  • Add to Cart