ENA333558420 Enamine

2-(4-aminophenyl)-2-methylpropanoic acid

  • MDL number MFCD13184255
  • Cas Number : 36402-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82328

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82328
Availability details for
Quantity
  • Availability
  • Add to Cart