ENA333558427 Enamine

4-(5-fluoro-1-methyl-1H-1,3-benzodiazol-2-yl)-2-methylaniline

  • MDL number MFCD12603548
  • Cas Number : 1182902-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-82338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82338
Availability details for
Quantity
  • Availability
  • Add to Cart