ENA333558430 Enamine

1-{[(tert-butoxy)carbonyl]amino}-2-methylcyclohexane-1-carboxylic acid

  • MDL number MFCD13502928
  • Cas Number : 1292818-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82575
Availability details for
Quantity
  • Availability
  • Add to Cart