ENA333558438 Enamine

3-(5-bromofuran-2-yl)-1,2-oxazol-5-amine

  • MDL number MFCD09703132
  • Cas Number : 938122-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82341

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82341
Availability details for
Quantity
  • Availability
  • Add to Cart