ENA333558440 Enamine

2-methyl-2-phenylpentan-3-one

  • MDL number MFCD17288540
  • Cas Number : 828-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82346
Availability details for
Quantity
  • Availability
  • Add to Cart