ENA333558448 Enamine

1-[2-(dimethylamino)ethyl]-1H-indol-6-amine

  • MDL number MFCD11638193
  • Cas Number : 173386-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82596
Availability details for
Quantity
  • Availability
  • Add to Cart