ENA333558454 Enamine

2-[2-(dimethylamino)ethoxy]-4-methylaniline

  • MDL number MFCD08687604
  • Cas Number : 946774-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82603

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82603
Availability details for
Quantity
  • Availability
  • Add to Cart