ENA333558456 Enamine

3-(cyclohexyloxy)benzoic acid

  • MDL number MFCD11116366
  • Cas Number : 158861-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82607
Availability details for
Quantity
  • Availability
  • Add to Cart