ENA333558460 Enamine

2-amino-2-cyclopropylpropanoic acid hydrochloride

  • MDL number MFCD19982412
  • Cas Number : 88807-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82618

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82618
Availability details for
Quantity
  • Availability
  • Add to Cart