ENA333558461 Enamine

4-(aminomethyl)-1-methylpiperidin-4-ol

  • MDL number MFCD08144105
  • Cas Number : 26228-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82619
Availability details for
Quantity
  • Availability
  • Add to Cart