ENA333558463 Enamine

3-(hydroxymethyl)-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD09817634
  • Cas Number : 951625-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82621
Availability details for
Quantity
  • Availability
  • Add to Cart