ENA333558472 Enamine

2-[4-(1,3-thiazole-4-amido)phenyl]acetic acid

  • MDL number MFCD17616834
  • Cas Number : 1334146-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82632
Availability details for
Quantity
  • Availability
  • Add to Cart