ENA333558485 Enamine

4-(3-methylmorpholin-4-yl)aniline

  • MDL number MFCD12134220
  • Cas Number : 1156819-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82357
Availability details for
Quantity
  • Availability
  • Add to Cart