ENA333558490 Enamine

2-[(2-chlorophenyl)amino]-2-(2-fluorophenyl)acetic acid

  • MDL number MFCD12899157
  • Cas Number : 1218090-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82661

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82661
Availability details for
Quantity
  • Availability
  • Add to Cart