ENA333558492 Enamine

(diamino-1,3,5-triazin-2-yl)methanol

  • MDL number MFCD12138109

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-82663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82663
Availability details for
Quantity
  • Availability
  • Add to Cart