ENA333558493 Enamine

(1-benzylpiperidin-4-yl)urea

  • MDL number MFCD09803352
  • Cas Number : 61220-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82664
Availability details for
Quantity
  • Availability
  • Add to Cart