ENA333558496 Enamine

2-benzyl-3-methylbutanoic acid

  • MDL number MFCD12794598
  • Cas Number : 53483-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82667

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82667
Availability details for
Quantity
  • Availability
  • Add to Cart