ENA333558505 Enamine

(1-benzoylpiperidin-4-yl)methanamine

  • MDL number MFCD10016285
  • Cas Number : 262864-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82366

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82366
Availability details for
Quantity
  • Availability
  • Add to Cart