ENA333558513 Enamine

1-(1,3-dioxaindan-5-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11193607
  • Cas Number : 1152534-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82686

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82686
Availability details for
Quantity
  • Availability
  • Add to Cart