ENA333558523 Enamine

2-(3-fluorophenyl)-2-methylpropanenitrile

  • MDL number MFCD11036565
  • Cas Number : 93748-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82384
Availability details for
Quantity
  • Availability
  • Add to Cart