ENA333558529 Enamine

methyl 3-amino-3-(4-tert-butylphenyl)propanoate hydrochloride

  • MDL number MFCD19982392
  • Cas Number : 1333871-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82392
Availability details for
Quantity
  • Availability
  • Add to Cart