ENA333558567 Enamine

4-cyano-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD16093711
  • Cas Number : 1251110-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82758

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-82758
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart