ENA333558574 Enamine

7-{[(benzyloxy)carbonyl]amino}heptanoic acid

  • MDL number MFCD09930580
  • Cas Number : 23434-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82766
Availability details for
Quantity
  • Availability
  • Add to Cart