ENA333558594 Enamine

4-methylbenzoyl isocyanate

  • MDL number MFCD09813452
  • Cas Number : 5843-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82406
Availability details for
Quantity
  • Availability
  • Add to Cart