ENA333558601 Enamine

1-(3-bromofuran-2-yl)ethan-1-one

  • MDL number MFCD19611108
  • Cas Number : 22037-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-82813
Availability details for
Quantity
  • Availability
  • Add to Cart