ENA333558622 Enamine

{2-[2-(dimethylamino)ethoxy]pyridin-3-yl}methanamine

  • MDL number MFCD12786418
  • Cas Number : 1249494-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82837
Availability details for
Quantity
  • Availability
  • Add to Cart