ENA333558629 Enamine

4-(2-phenoxyethoxy)piperidine hydrochloride

  • MDL number MFCD13560982
  • Cas Number : 1219980-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82846
Availability details for
Quantity
  • Availability
  • Add to Cart