ENA333558630 Enamine

N-ethyl-1-[6-methyl-2-(propan-2-yl)pyrimidin-4-yl]piperidin-4-amine

  • MDL number MFCD12605904
  • Cas Number : 1179313-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-82867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82867
Availability details for
Quantity
  • Availability
  • Add to Cart